Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016247.3 | NP_057331.2 | 9 - 10 | 1114 | 102 | |
GenBank mRNA | AB593127.1 | - | 9 - 10 | 1105 | 102 | |
AF157624.1 | - | 9 - 10 | 925 | 102 | ||
AF173155.1 | - | 9 - 10 | 1102 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons |