Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028999.hg.1 |
TC02001807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028999.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028999.hg.1 |
ENST00000490133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028999.hg.1 |
ENST00000535761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02028999.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000461805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000475073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
ENST00000493649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012459.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
ENST00000263736.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
ENST00000490133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
NM_018079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200190967.hg.1 |
TC0200012459.hg.1 |
Clariom D TC |