Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034370.hg.1 |
TC03001907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034370.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034365.hg.1 |
TC03001907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034365.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034365.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034365.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034367.hg.1 |
TC03001907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034367.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034367.hg.1 |
ENST00000455408.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034367.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034367.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195930.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195930.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195930.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195930.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195930.hg.1 |
TC0300012816.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012816.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
ENST00000302632.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
ENST00000468596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
NM_022788.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
NM_176876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195927.hg.1 |
TC0300012816.hg.1 |
Clariom D TC |