Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111989.1 | 1 - 2 | 192 | 64 | ||
GenBank mRNA | AK024529.1 | - | 1 - 2 | 161 | 64 | |
AK225030.1 | - | 1 - 2 | 161 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons |