Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
TC06001955.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
ENST00000229955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
ENST00000369268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
ENST00000417980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
ENST00000536583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024064.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
TC06001955.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
ENST00000229955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
ENST00000369268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
ENST00000417980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
ENST00000536583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024062.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
TC06001955.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
ENST00000229955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
ENST00000417980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
ENST00000536583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024067.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
TC06001955.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
ENST00000229955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
ENST00000369268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
ENST00000417980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
ENST00000536583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024061.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186967.hg.1 |
TC0600012625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186968.hg.1 |
TC0600012625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186970.hg.1 |
TC0600012625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186972.hg.1 |
TC0600012625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
ENST00000229955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_001143957.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012625.hg.1 |
NM_030784.3 |
Refseq |