Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
TC06001328.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
ENST00000378119.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
ENST00000465685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
ENST00000540769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06015910.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600174914.hg.1 |
TC0600011071.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378102.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
ENST00000378119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011071.hg.1 |
NM_030939.4 |
Refseq |