Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008604.hg.1 |
ENST00000224807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008604.hg.1 |
ENST00000393459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008604.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008604.hg.1 |
TC10000731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008603.hg.1 |
ENST00000224807.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008603.hg.1 |
ENST00000393459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008603.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008603.hg.1 |
TC10000731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000465383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000466982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000470252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000487721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008668.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159205.hg.1 |
TC1000008668.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000224807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000393459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
ENST00000489434.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
NM_030971.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159207.hg.1 |
TC1000008668.hg.1 |
Clariom D TC |