Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000337919.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000354724.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000435063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000435063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000435063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000435063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000435063.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000518733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000518733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000518733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000518733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000518733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000519075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000521111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
ENST00000523976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001040708.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_001282851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010894.hg.1 |
NM_012258.3 |
Refseq |