Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024556.hg.1 |
TC05001803.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024556.hg.1 |
ENST00000314744.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024556.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183785.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183785.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183785.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183785.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183785.hg.1 |
TC0500012105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500183784.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183784.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183784.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183784.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183784.hg.1 |
TC0500012105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
ENST00000314744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012105.hg.1 |
NM_006161.2 |
Refseq |