Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK023936.1 | - | 1 | 878 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |