Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006506.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006506.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006506.hg.1 |
TC20000468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006507.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006507.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006507.hg.1 |
TC20000468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006508.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006508.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006508.hg.1 |
TC20000468.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000155136.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155136.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155136.hg.1 |
TC2000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155136.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155136.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155135.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155135.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155135.hg.1 |
TC2000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155135.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155135.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155137.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155137.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155137.hg.1 |
TC2000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155137.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155137.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155134.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155134.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155134.hg.1 |
TC2000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155134.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155134.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
ENST00000217133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007925.hg.1 |
NM_030773.3 |
Refseq |