Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011395.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011395.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018466.hg.1 |
TC0X001373.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018466.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018466.hg.1 |
ENST00000406273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018466.hg.1 |
ENST00000511190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018466.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011395.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011395.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011395.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179035.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179035.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179035.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179035.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179035.hg.1 |
TC0X00011395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00179036.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179036.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179036.hg.1 |
ENST00000370832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00179036.hg.1 |
NM_031907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00179036.hg.1 |
TC0X00011395.hg.1 |
Clariom D TC |