Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000306422.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000515716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
ENST00000515716.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011211.hg.1 |
NM_032109.2 |
Refseq |