Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
TC09000663.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
ENST00000314904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
ENST00000373387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
ENST00000373399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
ENST00000435213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007739.hg.1 |
NM_032293.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
TC09000663.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
ENST00000314904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
ENST00000373387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
ENST00000373399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
ENST00000435213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007740.hg.1 |
NM_032293.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000373386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000373387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000435213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000495172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NM_001286779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NM_032293.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NR_104591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000373386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000373387.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000435213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
ENST00000495172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NM_001286779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NM_032293.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
NR_104591.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184668.hg.1 |
TC0900012173.hg.1 |
Clariom D TC |