Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010733.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010733.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010733.hg.1 |
TC11000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008550.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008550.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008550.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008550.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163310.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163310.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163310.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163310.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163310.hg.1 |
TC1100008550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008550.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008550.hg.1 |
ENST00000281030.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008550.hg.1 |
NM_003251.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008550.hg.1 |
NM_003251.3 |
Refseq |