Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010184.hg.1 |
TC01000600.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010184.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010184.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158680.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158680.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158680.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158680.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158680.hg.1 |
TC0100008194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100158681.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158681.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158681.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100158681.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100158681.hg.1 |
TC0100008194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
ENST00000334793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008194.hg.1 |
NM_004474.3 |
Refseq |