Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
TC06000385.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
ENST00000375650.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
ENST00000542758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
ENST00000545241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
ENST00000545429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003977.hg.1 |
NM_005346.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001225.hg.1 |
TC6_qbl_hap6000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001225.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
TC06000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
ENST00000375651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
ENST00000375652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
ENST00000458062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
ENST00000541556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003951.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
TC06000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
ENST00000375651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
ENST00000375652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
ENST00000435626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
ENST00000458062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
ENST00000541556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003953.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000499.hg.1 |
TC6_apd_hap1000039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000499.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001213.hg.1 |
TC6_dbb_hap3000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001213.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
TC06000385.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
ENST00000375650.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
ENST00000418382.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
ENST00000542758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
ENST00000545241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
ENST00000545429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003976.hg.1 |
NM_005346.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001270.hg.1 |
TC6_cox_hap2000078.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001270.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
TC06000385.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
ENST00000375650.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
ENST00000542758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
ENST00000545241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
ENST00000545429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003975.hg.1 |
NM_005346.4 |
Refseq |