Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007993.hg.1 |
ENST00000371021.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007993.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007993.hg.1 |
TC10000692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007994.hg.1 |
ENST00000371021.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007994.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007994.hg.1 |
TC10000692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007995.hg.1 |
ENST00000371021.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007995.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007995.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007995.hg.1 |
TC10000692.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000158386.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158386.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158386.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158386.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158386.hg.1 |
TC1000008568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158389.hg.1 |
TC1000008568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158387.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158387.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158387.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158387.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158387.hg.1 |
TC1000008568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
ENST00000490980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008568.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
ENST00000371019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011546.hg.1 |
NM_012083.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158385.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158385.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158385.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158385.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158385.hg.1 |
TC1000008568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158388.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158388.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158388.hg.1 |
ENST00000371021.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158388.hg.1 |
NM_005479.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158388.hg.1 |
TC1000008568.hg.1 |
Clariom D TC |