Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027386.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027386.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027386.hg.1 |
TC12002079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027387.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027387.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027387.hg.1 |
TC12002079.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
ENST00000328880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012247.hg.1 |
NM_177551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012248.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195892.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195892.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195892.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195892.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195892.hg.1 |
TC1200012248.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195891.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195891.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195891.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195891.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195891.hg.1 |
TC1200012248.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200195890.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195890.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195890.hg.1 |
ENST00000528880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200195890.hg.1 |
NM_006018.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200195890.hg.1 |
TC1200012248.hg.1 |
Clariom D TC |