Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017700.hg.1 |
ENST00000292609.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017700.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017700.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017700.hg.1 |
TC19001257.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
TC1900009873.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181032.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000292609.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000340880.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000594637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
ENST00000601792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009873.hg.1 |
NM_052890.3 |
Refseq |