Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127208.2 | NP_001120680.1 | 1 - 2 | 300 | 68 | |
XM_006714242.3 | XP_006714305.1 | 1 - 2 | 144 | 68 | ||
XM_017008319.1 | XP_016863808.1 | 1 - 2 | 144 | 68 | ||
GenBank mRNA | AI084057.1 | - | 1 - 2 | 68 | ||
AK055149.1 | - | 1 - 2 | 107 | 68 | ||
DA391215.1 | - | 1 - 2 | 109 | 68 | ||
FM992369.1 | - | 1 - 2 | 692 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons |