Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SAYSVFN motif domain containing 1 | SAYSD1 | C6orf64 | -23.4kb | Sense | Chr. 6 - 39104063 - 39115186 on NCBI Build GRCh38 |
Downstream | potassium two pore domain channel subfamily K member 5 | KCNK5 |
K2p5.1, KCNK5b, TASK-2, TASK2 |
48.4kb | Sense | Chr. 6 - 39188971 - 39229475 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |