Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003981.hg.1 |
ENST00000241051.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003981.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003981.hg.1 |
TC11000312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003984.hg.1 |
ENST00000241051.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003984.hg.1 |
ENST00000311388.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003984.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003984.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003984.hg.1 |
TC11000312.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150821.hg.1 |
TC1100007220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150826.hg.1 |
TC1100007220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000311388.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
ENST00000616885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007220.hg.1 |
NM_139160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150820.hg.1 |
TC1100007220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000241051.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000427755.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
ENST00000532078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
NM_001077242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150822.hg.1 |
TC1100007220.hg.1 |
Clariom D TC |