Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001189.hg.1 |
ENST00000333751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001189.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001189.hg.1 |
TC17000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001188.hg.1 |
ENST00000333751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001188.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001188.hg.1 |
TC17000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001191.hg.1 |
ENST00000333751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001191.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001191.hg.1 |
TC17000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001190.hg.1 |
ENST00000333751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001190.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001190.hg.1 |
TC17000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146829.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146829.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146829.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146829.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146829.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146822.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146822.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146822.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146822.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146822.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146827.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146827.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146827.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146827.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146827.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146826.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146826.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146826.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146826.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146826.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146828.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146828.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146828.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146828.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146828.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146823.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146824.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146825.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146825.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146825.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146825.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146825.hg.1 |
TC1700006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000333751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
ENST00000576980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006741.hg.1 |
NM_001002914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009632.hg.1 |
ENST00000572417.1 |
Ensembl |