Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027643.hg.1 |
TC07001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027643.hg.1 |
ENST00000297145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027643.hg.1 |
ENST00000358032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027643.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027642.hg.1 |
TC07001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027642.hg.1 |
ENST00000297145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027642.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027647.hg.1 |
TC07001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027647.hg.1 |
ENST00000297145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027647.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027644.hg.1 |
TC07001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027644.hg.1 |
ENST00000297145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027644.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027637.hg.1 |
TC07001765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027637.hg.1 |
ENST00000297145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027637.hg.1 |
ENST00000358032.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027637.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027637.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188676.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188676.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188676.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188676.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188676.hg.1 |
TC0700012296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700188677.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188677.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188677.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188677.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188677.hg.1 |
TC0700012296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188675.hg.1 |
TC0700012296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188672.hg.1 |
TC0700012296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000297145.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000432572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
ENST00000485446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012296.hg.1 |
NM_152556.2 |
Refseq |