Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000370634.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000440515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000456412.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
ENST00000470358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008534.hg.1 |
NM_016267.3 |
Refseq |