Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127487.1 | NP_001120959.1 | 29 - 30 | 5159 | 70 | |
NM_001458.4 | NP_001449.3 | 29 - 30 | 5159 | 70 | ||
GenBank mRNA | AB208865.1 | - | 6 - 7 | 801 | 70 | |
AB371585.1 | - | 29 - 30 | 5179 | 70 | ||
AF089841.1 | - | 29 - 30 | 4999 | 70 | ||
AF146692.1 | - | 29 - 30 | 5162 | 70 | ||
AJ012737.1 | - | 29 - 30 | 4968 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |