Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288833.1 | NP_001275762.1 | 8 - 9 | 1374 | 145 | |
NM_013421.2 | NP_038265.2 | 9 - 10 | 1302 | 145 | ||
NM_013430.2 | NP_038347.2 | 8 - 9 | 1062 | 145 | ||
GenBank mRNA | BC025927.1 | - | 6 - 7 | 942 | 4548861 | 145 |
CR456494.1 | - | 5 - 6 | 652 | 145 | ||
J04131.1 | - | 9 - 10 | 1244 | 145 | ||
J05235.1 | - | 7 - 8 | 1319 | 145 | ||
M24087.1 | - | 4 - 5 | 577 | 145 | ||
M24903.1 | - | 8 - 9 | 1062 | 145 | ||
X60069.1 | - | 6 - 7 | 933 | 145 |
Set Membership: |
Probe spans exons |