Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000264.hg.1 |
TC06000032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000264.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000264.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000264.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000267.hg.1 |
TC06000032.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000267.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000267.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000267.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
ENST00000380272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
ENST00000380274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
NM_012135.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600146172.hg.1 |
TC0600006714.hg.1 |
Clariom D TC |