Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030662.3 | NP_109587.1 | 6 - 7 | 961 | 61 | |
XM_017026989.1 | XP_016882478.1 | 6 - 7 | 1016 | 61 | ||
GenBank mRNA | AA312322.1 | - | 4 - 5 | 405 | 61 | |
AK093125.1 | - | 3 - 4 | 1008 | 61 | ||
BC000471.2 | - | 6 - 7 | 961 | 2961198 | 61 | |
BC018645.2 | - | 6 - 7 | 903 | 4025987 | 61 | |
BX538081.1 | - | 7 - 8 | 711 | 61 | ||
L11285.1 | - | 6 - 7 | 791 | 61 |
Set Membership: |
Detect Genomic DNA Probe spans exons |