Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
TC08001733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
ENST00000349157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
ENST00000526683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
ENST00000527197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022883.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022880.hg.1 |
TC08001733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022880.hg.1 |
ENST00000349157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022880.hg.1 |
ENST00000526683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022880.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022880.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
TC08001733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000313352.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000349157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000453551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000456095.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000524570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000526683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
ENST00000527197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
NM_001136033.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022873.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
TC08001733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
ENST00000313352.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
ENST00000349157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
ENST00000453551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
ENST00000526683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
NM_001136033.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022875.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000313352.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000349157.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000453551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000527584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000527744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000528320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000529999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000531897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000531951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000532127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000533162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000533362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001136033.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000313352.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000349157.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000453551.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526459.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000526683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000527584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000527744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000528320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000529999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000531897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000531951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000532127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000533162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
ENST00000533362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001136033.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271098.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_001271100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_014281.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
NM_078480.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183094.hg.1 |
TC0800012167.hg.1 |
Clariom D TC |