Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016391.hg.1 |
ENST00000324767.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016391.hg.1 |
ENST00000538941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016391.hg.1 |
NM_001145923.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016391.hg.1 |
NM_020843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016391.hg.1 |
TC15001688.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000324767.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000538941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000562948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000563246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000563290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
NM_001145923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
NM_020843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000324767.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000538941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000562948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000563246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
ENST00000563290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
NM_001145923.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
NM_020843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175568.hg.1 |
TC1500010097.hg.1 |
Clariom D TC |