Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034311.hg.1 |
TC03003420.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034311.hg.1 |
ENST00000295911.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034311.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
TC03003419.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
TC03003420.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
ENST00000295911.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
ENST00000327047.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
ENST00000328863.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034309.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034312.hg.1 |
TC03003420.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034312.hg.1 |
ENST00000295911.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034312.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209334.hg.1 |
TC0300014069.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014069.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209332.hg.1 |
TC0300014069.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000295911.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000327047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000328863.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000468836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000472224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
ENST00000485607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_001195794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_001256819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_052995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NM_174878.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
NR_046380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209330.hg.1 |
TC0300014069.hg.1 |
Clariom D TC |