Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
TC01003359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
ENST00000368192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052521.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052525.hg.1 |
TC01003359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052525.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052525.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052526.hg.1 |
TC01003359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052526.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052523.hg.1 |
TC01003359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052523.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052523.hg.1 |
ENST00000368192.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052523.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052523.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221770.hg.1 |
TC0100016035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016035.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221773.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221773.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221773.hg.1 |
TC0100016035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000368192.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
ENST00000460850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
NM_005240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221767.hg.1 |
TC0100016035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100221772.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221772.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221772.hg.1 |
ENST00000326786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221772.hg.1 |
NM_001145312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221772.hg.1 |
TC0100016035.hg.1 |
Clariom D TC |