Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314044.1 | NP_001300973.1 | 5 - 6 | 538 | 133 | |
NM_001314045.1 | NP_001300974.1 | 5 - 6 | 546 | 133 | ||
NM_001314046.1 | NP_001300975.1 | 5 - 6 | 533 | 133 | ||
NM_001314047.1 | NP_001300976.1 | 5 - 6 | 520 | 133 | ||
NM_001314048.1 | NP_001300977.1 | 4 - 5 | 425 | 133 | ||
NM_033439.3 | NP_254274.1 | 5 - 6 | 551 | 133 | ||
XM_011518061.2 | XP_011516363.1 | 4 - 5 | 486 | 133 | ||
XM_017015285.1 | XP_016870774.1 | 7 - 8 | 1279 | 133 | ||
GenBank mRNA | AB024518.1 | - | 4 - 5 | 490 | 133 | |
AY905581.1 | - | 4 - 5 | 473 | 133 | ||
BC047085.1 | - | 5 - 6 | 528 | 5314040 | 133 | |
BQ878541.1 | - | 4 - 5 | 512 | 133 | ||
CR407619.1 | - | 4 - 5 | 473 | 133 | ||
DB266668.1 | - | 5 - 6 | 512 | 133 | ||
HQ641439.1 | - | 3 - 4 | 362 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |