Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000274487.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000502709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000505791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
ENST00000509467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008586.hg.1 |
NM_133638.4 |
Refseq |