Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001894.hg.1 |
TC0Y000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001894.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001894.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007188.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148324.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148324.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148324.hg.1 |
ENST00000330337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148324.hg.1 |
NR_001537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148324.hg.1 |
TC0Y00007188.hg.1 |
Clariom D TC |