Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
ENST00000269051.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
ENST00000431505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
ENST00000536287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
ENST00000538145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004733.hg.1 |
TC17000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700201778.hg.1 |
TC1700012229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000269051.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000431505.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000536287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000538145.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000578006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
ENST00000582967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012229.hg.1 |
NM_138328.2 |
Refseq |