Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006901.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006901.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006901.hg.1 |
TC11000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006900.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006900.hg.1 |
ENST00000310969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006900.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006900.hg.1 |
TC11000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006899.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006899.hg.1 |
ENST00000310969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006899.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006899.hg.1 |
TC11000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006902.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006902.hg.1 |
ENST00000310969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006902.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006902.hg.1 |
TC11000570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006898.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006898.hg.1 |
ENST00000310969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006898.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006898.hg.1 |
TC11000570.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156273.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156274.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156274.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156274.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156274.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156274.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156270.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156271.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156272.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156275.hg.1 |
TC1100007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000279178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
ENST00000536333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007892.hg.1 |
NM_080866.2 |
Refseq |