Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009765.hg.1 |
ENST00000323346.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009765.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009765.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009765.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12009765.hg.1 |
TC12000776.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000547922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
ENST00000552697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008593.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
ENST00000323346.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
ENST00000392989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
NM_001145288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
NM_139319.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200163173.hg.1 |
TC1200008593.hg.1 |
Clariom D TC |