Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000522278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
ENST00000522278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013319.hg.1 |
TC08001045.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013319.hg.1 |
ENST00000256404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013319.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000521284.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000522278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
ENST00000522278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009858.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166936.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166936.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166936.hg.1 |
ENST00000256404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166936.hg.1 |
NM_144962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166936.hg.1 |
TC0800009858.hg.1 |
Clariom D TC |