Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
TC08000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000284503.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000382309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000403422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000436750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000455213.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
ENST00000528323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
NM_001135746.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
NM_001135747.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
NM_001135748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000888.hg.1 |
NM_145043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000284503.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000403422.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000436750.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000455213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000524741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000528113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000528323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135746.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135747.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_145043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000284503.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000403422.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000436750.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000455213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000524741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000528113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
ENST00000528323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135746.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135747.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_001135748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
NM_145043.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145796.hg.1 |
TC0800006758.hg.1 |
Clariom D TC |