Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000346.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000346.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000346.hg.1 |
TC14000070.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000344.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000344.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000344.hg.1 |
TC14000070.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400144615.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144615.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144615.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144615.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144615.hg.1 |
TC1400006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144613.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144613.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144613.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144613.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144613.hg.1 |
TC1400006534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
ENST00000304677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006534.hg.1 |
NM_005615.4 |
Refseq |