Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007102.hg.1 |
ENST00000246841.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007102.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007102.hg.1 |
TC11000584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007103.hg.1 |
ENST00000246841.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007103.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007103.hg.1 |
TC11000584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007101.hg.1 |
ENST00000246841.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007101.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007101.hg.1 |
TC11000584.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100201007.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201007.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201007.hg.1 |
TC1100013030.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100201005.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201005.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201005.hg.1 |
TC1100013030.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100201006.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201006.hg.1 |
NM_013280.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100201006.hg.1 |
TC1100013030.hg.1 |
Clariom D TC |