Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC009851.2 | - | 7 - 8 | 1719 | 94 | |
BC011857.2 | - | 8 - 9 | 1738 | 94 | ||
BC017356.2 | - | 7 - 8 | 1755 | 94 | ||
BC019235.2 | - | 7 - 8 | 1757 | 95 | ||
BC020240.1 | - | 7 - 8 | 1786 | 94 | ||
BC089412.1 | - | 6 - 7 | 1752 | 94 | ||
X17115.1 | - | 8 - 9 | 1830 | 94 | ||
X58529.1 | - | 5 - 6 | 1943 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |