Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042652.hg.1 |
TC01002718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042652.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042652.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000450089.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000486478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000488027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
ENST00000491817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014368.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207621.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207621.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207621.hg.1 |
ENST00000371201.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207621.hg.1 |
NM_152377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207621.hg.1 |
TC0100014368.hg.1 |
Clariom D TC |