Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002576.hg.1 |
TC6_cox_hap2000150.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002576.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002507.hg.1 |
TC6_dbb_hap3000139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002507.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002095.hg.1 |
TC6_mann_hap4000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002095.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002238.hg.1 |
TC6_mcf_hap5000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002238.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002243.hg.1 |
TC6_mcf_hap5000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002243.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017163.hg.1 |
TC06001518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017163.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017163.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002091.hg.1 |
TC6_mann_hap4000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002091.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002239.hg.1 |
TC6_mcf_hap5000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002239.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002547.hg.1 |
TC6_qbl_hap6000140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002547.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002550.hg.1 |
TC6_qbl_hap6000140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002550.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002090.hg.1 |
TC6_mann_hap4000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002090.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017166.hg.1 |
TC06001518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017166.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017166.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002508.hg.1 |
TC6_dbb_hap3000139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002508.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002510.hg.1 |
TC6_dbb_hap3000139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002510.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000493289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000493289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017164.hg.1 |
TC06001518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017164.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017164.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002575.hg.1 |
TC6_cox_hap2000150.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002575.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002512.hg.1 |
TC6_dbb_hap3000139.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002512.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002093.hg.1 |
TC6_mann_hap4000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002093.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002552.hg.1 |
TC6_qbl_hap6000140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002552.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002241.hg.1 |
TC6_mcf_hap5000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002241.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002548.hg.1 |
TC6_qbl_hap6000140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002548.hg.1 |
NM_001264.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176905.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176905.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176905.hg.1 |
TC0600011402.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000259881.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000467107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000479581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000481450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000493289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000493289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000547221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000548049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000550838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007562.hg.1 |
ENST00000552747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011402.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176904.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176904.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176904.hg.1 |
TC0600011402.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600176906.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176906.hg.1 |
ENST00000376288.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176906.hg.1 |
TC0600011402.hg.1 |
Clariom D TC |