Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565781 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565835 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705143 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565809 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565814 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705066 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565802 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565838 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705071 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565798 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565828 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565767 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565812 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565787 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565827 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15485340 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565805 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705095 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705105 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705082 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565801 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565791 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705123 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565771 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565831 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705084 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565788 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565761 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565836 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705067 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565795 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565774 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565764 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565834 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565777 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705077 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565822 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565821 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705116 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565813 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565766 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565773 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565824 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705141 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565768 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15485321 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565808 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565818 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565760 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565830 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565833 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565841 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565779 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705087 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22705075 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565825 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15485333 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15485328 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02565820 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
176264 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
60798 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
102582 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
468124 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
929 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
647388 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
937965 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
AF132973.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
AF155662.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
AF261134.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
AF286697.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
BC000589.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
BC009189.2 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
150996 |
NM_015965.5 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
499568 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P395585 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P395585 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
3897 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P103045 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P103045 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
57963 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31097805 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31097786 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P58554742 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P395585 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P395585 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565799 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565802 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565806 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565825 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565784 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565820 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565813 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565796 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565795 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565801 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705143 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565793 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705141 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565805 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565831 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565788 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565808 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565828 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565814 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565818 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565810 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15485333 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565791 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565798 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705123 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565833 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705105 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565822 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565823 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705142 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705141 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565810 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565818 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705143 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705142 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565820 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565828 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565813 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565801 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705105 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565823 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565799 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565793 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565833 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565791 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565814 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565796 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565808 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565806 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565789 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565825 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565805 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565784 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565788 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565798 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565795 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15485333 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565802 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22705123 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565831 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02565822 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P395585 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P103045 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565818 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565823 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565828 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565812 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565820 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565825 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565831 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565833 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02565814 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00805813 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P103045 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P103045 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P395585 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
78238 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
19428 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16395066 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16395071 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P103045 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00805818 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31097805 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P23156753 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14378527 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00805795 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565802 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565833 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565776 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565778 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565784 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565820 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565765 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565825 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565796 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565781 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565728 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565763 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565793 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565799 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565768 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565828 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565840 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565788 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565809 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15485319 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565772 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565804 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565760 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15485335 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15485321 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565831 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565773 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565761 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565818 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565838 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565814 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565783 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565842 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565789 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565792 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565795 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565786 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565764 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565801 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15485323 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565758 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565811 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565770 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15485339 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565798 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02565823 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16395066 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P103045 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P23156753 |
- |
- |