Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
TC05001678.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
ENST00000379638.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
ENST00000513339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
ENST00000545426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022756.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181045.hg.1 |
TC0500011721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000261482.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000379638.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000474542.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000497856.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000504247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000511865.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
ENST00000513339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011721.hg.1 |
NM_005669.4 |
Refseq |