Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018221.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018221.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018221.hg.1 |
TC16001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018218.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018218.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018218.hg.1 |
TC16001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018220.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018220.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018220.hg.1 |
TC16001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007229.hg.1 |
ENST00000455360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018219.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018219.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018219.hg.1 |
TC16001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007034.hg.1 |
ENST00000422309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007229.hg.1 |
ENST00000455360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186010.hg.1 |
TC1600011060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000561707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011060.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007034.hg.1 |
ENST00000422309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
ENST00000567786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186006.hg.1 |
TC1600011060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186009.hg.1 |
TC1600011060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000262428.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000564057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
ENST00000564662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
NM_021149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186011.hg.1 |
TC1600011060.hg.1 |
Clariom D TC |